(4-Hydroxybenzyl)phosphonic acid ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
H1558-200MG
H1558-1G
TCIAH1558-200MGEA
510
SEK
TCIAH1558-200MG
TCIAH1558-1G
(4-Hydroxybenzyl)phosphonic acid ≥98.0%
(4-Hydroxybenzyl)phosphonic acid
Formel:
C₇H₉O₄P MW: 188,12 g/mol |
MDL Number:
MFCD16877729 CAS nummer: 90001-07-3 UN: 3261 ADR: 8,III |
Specifikation testresultat
Appearance | White to Almost white powder to crystaline |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 220.0 to 224.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...