Hexylphosphonic acid ≥98.0%

Leverantör: TCI
Fara

H1551-1G H1551-5G
TCIAH1551-1GEA 685 SEK
TCIAH1551-1G TCIAH1551-5G
Hexylphosphonic acid ≥98.0%
Hexylphosphonic acid
Formel: C₆H₁₅O₃P
MDL Number: MFCD00013961
CAS nummer: 4721-24-8
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cdd47a940e89741a62c8f5086d50c503862b83c&vid=7cdd47a940e89741a62c8f5086d50c503862b83c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 107.0 to 111.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR