5-(4-Hydroxypiperidino)-2-thiophenecarbaldehyde ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
5-(4-Hydroxypiperidin-1-yl)thiophene-2-carbaldehyde, 5-(4-Hydroxypiperidin-1-yl)thiophene-2-carboxaldehyde, 5-(4-Hydroxypiperidino)thiophene-2-carboxaldehyde
H1550-200MG
H1550-1G
TCIAH1550-200MGEA
215
SEK
TCIAH1550-200MG
TCIAH1550-1G
5-(4-Hydroxypiperidino)-2-thiophenecarbaldehyde ≥98.0%
5-(4-Hydroxypiperidino)-2-thiophenecarbaldehyde
Formel:
C₁₀H₁₃NO₂S MW: 211,28 g/mol |
MDL Number:
MFCD03425728 CAS nummer: 207290-72-0 |
Specifikation testresultat
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 143.0 to 147.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...