2-(Hydroxymethyl)-4-nitrobenzenol ≥98.0%

Leverantör: TCI

H1549-5G H1549-25G
TCIAH1549-5GEA 1500 SEK
TCIAH1549-5G TCIAH1549-25G
2-(Hydroxymethyl)-4-nitrobenzenol ≥98.0%
2-(Hydroxymethyl)-4-nitrobenzenol
Formel: C₇H₇NO₄
MDL Number: MFCD01321361
CAS nummer: 39224-61-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3da7f24ebf72d831ab7c6bddbc05b4e7bc208ff9&vid=3da7f24ebf72d831ab7c6bddbc05b4e7bc208ff9&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 123.0 to 127.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR