2-(1-Hydroxy-3-methylbutylidene)-5,5-dimethyl-1,3-cyclohexanedione ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 2-(1-Hydroxy-3-methylbutylidene)-5,5-dimethylcyclohexane-1,3-dione, 2-(1-Hydroxy-3-methylbutylidene)dimedone
H1546-1G H1546-5G
TCIAH1546-1GEA 790 SEK
TCIAH1546-1G TCIAH1546-5G
2-(1-Hydroxy-3-methylbutylidene)-5,5-dimethyl-1,3-cyclohexanedione ≥98.0%
2-(1-Hydroxy-3-methylbutylidene)-5,5-dimethyl-1,3-cyclohexanedione
Formel: C₁₃H₂₀O₃
MW: 224,30 g/mol
MDL Number: MFCD01862890
CAS nummer: 172611-72-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=315df4e96d973c5cabf30855dc43a66c5ef27b25&vid=315df4e96d973c5cabf30855dc43a66c5ef27b25&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %