3-Hydroxybenzophenone ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
m-Hydroxybenzophenone, (3-Hydroxyphenyl)(phenyl)methanone
H1518-5G
H1518-25G
TCIAH1518-5GEA
1500
SEK
TCIAH1518-5G
TCIAH1518-25G
3-Hydroxybenzophenone ≥98.0%
3-Hydroxybenzophenone
Formel:
C₁₃H₁₀O₂ MW: 198,22 g/mol |
MDL Number:
MFCD00002297 CAS nummer: 13020-57-0 EINECS: 235-879-1 |
Specifikation testresultat
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 115.0 to 119.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...