6,6,7,7,8,8,8-Heptafluoro-5,5-bis(trifluoromethyl)-1,3-octadiene ≥98.0%

Leverantör: TCI EUROPE
Varning

H1515-5G H1515-25G
TCIAH1515-5GEA 1700 SEK
TCIAH1515-5G TCIAH1515-25G
6,6,7,7,8,8,8-Heptafluoro-5,5-bis(trifluoromethyl)-1,3-octadiene ≥98.0%
6,6,7,7,8,8,8-Heptafluoro-5,5-bis(trifluoromethyl)-1,3-octadiene
Formel: C₁₀H₅F₁₃
MW: 372,13 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD28014057
CAS nummer: 1429188-77-1
UN: 1993
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR