4-Hydroxybenzamidine hydrochloride ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-Hydroxyphenylamidine hydrochloride, 4-Hydroxybenzene-1-carboximidamide hydrochloride
H1510-1G
H1510-5G
TCIAH1510-1GEA
325
SEK
TCIAH1510-1G
TCIAH1510-5G
4-Hydroxybenzamidine hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Hydroxybenzamidine hydrochloride
Formel:
C₇H₉ClN₂O MW: 172,61 g/mol |
MDL Number:
MFCD00136405 CAS nummer: 38148-63-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 221.0 to 225.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...