1,1,1-Trifluoroacetone cyanohydrin ≥96.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 1,1,1-Trifluoroacetone Cyanohydrin, 2-Cyano-1,1,1-trifluoro-2-propanol, 2-Hydroxy-2-(trifluoromethyl)propionitrile

H1508-1G H1508-5G
TCIAH1508-1GEA 565 SEK
TCIAH1508-1G TCIAH1508-5G
1,1,1-Trifluoroacetone cyanohydrin ≥96.0%
1,1,1-Trifluoroacetone cyanohydrin
Formel: C₄H₄F₃NO
MW: 139,08 g/mol
Kokpunkt: 139°C
Flampunkt: 60°C
MDL Number: MFCD00040885
CAS nummer: 335-08-0
UN: 3275
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54a3cc8c5ac34ad0d3ff1ed0d8ca87cbf0034105&vid=54a3cc8c5ac34ad0d3ff1ed0d8ca87cbf0034105&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR