3-(Hydroxymethyl)biphenyl ≥96.0%
Leverantör: TCI EUROPE
Synonyms:
Biphenyl-3-methanol, [1,1'-Biphenyl]-3-ylmethanol, 3-Phenylbenzyl Alcohol
H1481-1G
H1481-5G
TCIAH1481-1GEA
1125
SEK
TCIAH1481-1G
TCIAH1481-5G
3-(Hydroxymethyl)biphenyl ≥96.0%
3-(Hydroxymethyl)biphenyl
Formel:
C₁₃H₁₂O MW: 184,24 g/mol |
MDL Number:
MFCD08703631 CAS nummer: 69605-90-9 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 48.0 to 52.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...