3-Hexadecylthiophene ≥97.0%
Leverantör: TCI EUROPE
H1462-1G
H1462-5G
H1462-25G
TCIAH1462-1GEA
245
SEK
TCIAH1462-1G
TCIAH1462-5G
TCIAH1462-25G
3-Hexadecylthiophene ≥97.0%
3-Hexadecylthiophene
Formel:
C₂₀H₃₆S MW: 308,57 g/mol |
MDL Number:
MFCD07368973 CAS nummer: 119269-24-8 |
Specifikation testresultat
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 97.0 % |
Melting point | 31.0 to 35.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...