1,2,3,4,5,6-Hexa-O-[11-[4-(4-hexylphenylazo)phenoxy]undecanoyl]-D-mannitol 97,0 HPLC_ANALYSMETOD

Leverantör: TCI

H1452-1G H1452-5G
TCIAH1452-1GEA 3325 SEK
TCIAH1452-1G TCIAH1452-5G
1,2,3,4,5,6-Hexa-O-[11-[4-(4-hexylphenylazo)phenoxy]undecanoyl]-D-mannitol 97,0 HPLC_ANALYSMETOD
1,2,3,4,5,6-Hexa-O-[11-[4-(4-hexylphenylazo)phenoxy]undecanoyl]-D-mannitol
Formel: C₁₈₀H₂₅₄N₁₂O₁₈
MW: 2874.08 g/mol
CAS nummer: 1093077-77-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1025962a2a3c5ee93e01e21b69388d2173b247d6&vid=1025962a2a3c5ee93e01e21b69388d2173b247d6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystaline
Purity (HPLC) min. 97.0 area% (Multi-peak total)
Functionality test Suitability for Photochemically Reversible Liquefaction and Solidification


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR