2-Hydroxydimetridazole ≥98.0% (by GC, titration analysis)
Leverantör: TCI
H1439-1G
H1439-5G
TCIAH1439-1GEA
1125
SEK
TCIAH1439-1G
TCIAH1439-5G
2-Hydroxydimetridazole ≥98.0% (by GC, titration analysis)
2-Hydroxydimetridazole
Formel:
C₅H₇N₃O₃ |
MDL Number:
MFCD00159675 CAS nummer: 936-05-0 EINECS: 213-312-9 |
|
Specifikation testresultat
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 115.0 to 119.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...