3-Hydroxy-ɑ-methylbenzyl alcohol ≥98.0%

Leverantör: TCI
Varning

Synonyms: 3-(1-Hydroxyethyl)phenol, 1-(3-Hydroxyphenyl)ethanol

H1433-1G H1433-5G
TCIAH1433-1GEA 710 SEK
TCIAH1433-1G TCIAH1433-5G
3-Hydroxy-ɑ-methylbenzyl alcohol ≥98.0%
3-Hydroxy-alpha-methylbenzyl alcohol
Formel: C₈H₁₀O₂
MDL Number: MFCD00238621
CAS nummer: 2415-09-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d03eaae7413424427d49f1179e133ee3707bcff&vid=6d03eaae7413424427d49f1179e133ee3707bcff&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 117.0 to 121.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR