3-Hydroxy-1-adamantaneacetic acid ≥95.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 1-(Carboxymethyl)-3-hydroxyadamantane, 2-(3-Hydroxyadamantan-1-yl)acetic acid

H1414-1G H1414-5G
TCIAH1414-1GEA 725 SEK
TCIAH1414-1G TCIAH1414-5G
3-Hydroxy-1-adamantaneacetic acid ≥95.0% (by GC, titration analysis)
3-Hydroxy-1-adamantaneacetic acid
Formel: C₁₂H₁₈O₃
MW: 210,27 g/mol
MDL Number: MFCD00167797
CAS nummer: 17768-36-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e60c1929847e617dec29cc6527969ca346e14405&vid=e60c1929847e617dec29cc6527969ca346e14405&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystaline
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 123.0 to 128.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR