6-Hydroxy-5-nitronicotinic acid 97,0 GC, T
Leverantör: TCI
|
Varning
|
H1410-1G
H1410-5G
TCIAH1410-1GEA
870
SEK
TCIAH1410-1G
TCIAH1410-5G
6-Hydroxy-5-nitronicotinic acid 97,0 GC, T
6-Hydroxy-5-nitronicotinic acid
Formel:
C₆H₄N₂O₅ MW: 184.1100 g/mol Smältpunkt: 291°C |
MDL Number:
MFCD02180840 CAS nummer: 6635-31-0 |
|
Specifikation testresultat
Appearance | Light yellow to Yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...