3,5-Heptanedione ≥97.0%

Leverantör: TCI EUROPE
Varning

H1395-5G H1395-25G
TCIAH1395-5GEA 695 SEK
TCIAH1395-5G TCIAH1395-25G
3,5-Heptanedione ≥97.0%
3,5-Heptanedione
Formel: C₇H₁₂O₂
MW: 128,17 g/mol
Kokpunkt: 177°C
Flampunkt: 57°C
MDL Number: MFCD00015186
CAS nummer: 7424-54-6
EINECS: 231-054-5
UN: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21b7a7fc667050bfaf5cf61210e729ed6a18350f&vid=21b7a7fc667050bfaf5cf61210e729ed6a18350f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR