6-Hydroxybenzothiazole ≥96.0%

Leverantör: TCI

Synonyms: 6-Benzothiazolol

H1380-1G H1380-5G
TCIAH1380-1GEA 3850 SEK
TCIAH1380-1G TCIAH1380-5G
6-Hydroxybenzothiazole ≥96.0%
6-Hydroxybenzothiazole
Formel: C₇H₅NOS
MW: 151.1857 g/mol
MDL Number: MFCD00057883
CAS nummer: 13599-84-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=737978fc5f172f4d8e30978faa054dfccf411c1d&vid=737978fc5f172f4d8e30978faa054dfccf411c1d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 96.0 %
Melting point 186.0 to 190.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR