1,1,4,7,10,10-Hexamethyltriethylenetetramine ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
N1,N1'-(Ethane-1,2-diyl)bis(N1,N2,N2-trimethylethane-1,2-diamine)
H1371-5ML
H1371-25ML
TCIAH1371-5MLEA
1100
SEK
TCIAH1371-5ML
TCIAH1371-25ML
1,1,4,7,10,10-Hexamethyltriethylenetetramine ≥98.0%
1,1,4,7,10,10-Hexamethyltriethylenetetramine
Formel:
C₁₂H₃₀N₄ MW: 230,39 g/mol Kokpunkt: 130°C Flampunkt: 102°C |
MDL Number:
MFCD00025684 CAS nummer: 3083-10-1 EINECS: 221-382-7 UN: 2735 ADR: 8,II |
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...