(1-Methyl-1H-imidazol-2-yl)methanol ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: 1-Methyl-2-imidazolemethanol

H1370-1G H1370-5G
TCIAH1370-1GEA 590 SEK
TCIAH1370-1G TCIAH1370-5G
(1-Methyl-1H-imidazol-2-yl)methanol ≥98.0% (by GC, titration analysis)
(1-Methyl-1H-imidazol-2-yl)methanol
Formel: C₅H₈N₂O
MW: 112,13 g/mol
MDL Number: MFCD00964673
CAS nummer: 17334-08-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=057704f2f60742379e213a9dfabc6744ce960358&vid=057704f2f60742379e213a9dfabc6744ce960358&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 115.0 to 119.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR