2-Hexylthiophene ≥98.0%

Leverantör: TCI EUROPE

H1350-5G H1350-25G
TCIAH1350-5GEA 455 SEK
TCIAH1350-5G TCIAH1350-25G
2-Hexylthiophene ≥98.0%
2-Hexylthiophene
Formel: C₁₀H₁₆S
MW: 168,3 g/mol
Kokpunkt: 92°C
Flampunkt: 100°C
MDL Number: MFCD00022535
CAS nummer: 18794-77-9
EINECS: 242-579-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93b44aeed679d29de5071f04994fd5eadfde0fbe&vid=93b44aeed679d29de5071f04994fd5eadfde0fbe&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR