Hexyl propionate ≥98.0%

Leverantör: TCI

Synonyms: Propionic Acid Hexyl Ester

H1330-25ML H1330-500ML
TCIAH1330-25MLEA 245 SEK
TCIAH1330-25ML TCIAH1330-500ML
Hexyl propionate ≥98.0%
Hexyl propionate
Formel: C₉H₁₈O₂
Kokpunkt: 186°C
Flampunkt: 65°C
MDL Number: MFCD00085197
CAS nummer: 2445-76-3
EINECS: 219-495-1

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR