4-(4-Hydroxy-3-methoxyphenyl)-2-butanone ≥98.0%
Leverantör: TCI EUROPE
Synonyms: Zingerone, Vanillylacetone
H1314-25G H1314-100G
TCIAH1314-25GEA 485 SEK
TCIAH1314-25G TCIAH1314-100G
4-(4-Hydroxy-3-methoxyphenyl)-2-butanone ≥98.0%
4-(4-Hydroxy-3-methoxyphenyl)-2-butanone
Formel: C₁₁H₁₄O₃
MW: 194,23 g/mol
Kokpunkt: 148°C
Storage Temperature: Kylskåp
MDL Number: MFCD00048232
CAS nummer: 122-48-5
EINECS: 204-548-3
Merck Index: 14,10166
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d40cfba66e5e535191065aa3ea7d0e651fdfbb75&vid=d40cfba66e5e535191065aa3ea7d0e651fdfbb75&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 39.0 to 43.0 deg-C