Honokiol ≥95.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 3',5-Diallyl-2,4'-dihydroxybiphenyl, 3',5-Diallylbiphenyl-2,4'-diol, 5,3'-Diallyl-2,4'-dihydroxybiphenyl

H1309-1G H1309-200MG
TCIAH1309-1GEA 2075 SEK
TCIAH1309-1G TCIAH1309-200MG
Honokiol ≥95.0%
Honokiol
Formel: C₁₈H₁₈O₂
MW: 266,34 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00016674
CAS nummer: 35354-74-6
Merck Index: 14,04742
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc4dda67e694d191da625ebbf336a919399154bd&vid=bc4dda67e694d191da625ebbf336a919399154bd&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 83.0 to 87.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR