4-Hexyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
2-(4-Hexyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Hexyl-2-thiopheneboronic Acid Pinacol Ester, 2-(4-Hexylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
H1294-5G
H1294-1G
TCIAH1294-5GEA
4825
SEK
TCIAH1294-5G
TCIAH1294-1G
4-Hexyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥98.0%
4-Hexyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene
Formel:
C₁₆H₂₇BO₂S MW: 294,26 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD14708172 CAS nummer: 883742-29-8 |
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...