9-(2-Hydroxyethyl)adenine ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
6-Amino-9-(2-hydroxyethyl)purine
H1290-25G
H1290-5G
TCIAH1290-25GEA
2000
SEK
TCIAH1290-25G
TCIAH1290-5G
9-(2-Hydroxyethyl)adenine ≥98.0% (by HPLC, titration analysis)
9-(2-Hydroxyethyl)adenine
Formel:
C₇H₉N₅O MW: 179,18 g/mol |
MDL Number:
MFCD04220770 CAS nummer: 707-99-3 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 240.0 to 244.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...