2-[2-Hydroxy-5-[2-(methacryloyloxy)ethyl]phenyl]-2H-benzotriazole ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
H1261-25G
H1261-500G
TCIAH1261-25GEA
485
SEK
TCIAH1261-25G
TCIAH1261-500G
2-[2-Hydroxy-5-[2-(methacryloyloxy)ethyl]phenyl]-2H-benzotriazole ≥98.0%
2-[2-Hydroxy-5-[2-(methacryloyloxy)ethyl]phenyl]-2H-benzotriazole
Formel:
C₁₈H₁₇N₃O₃ MW: 323,35 g/mol |
MDL Number:
MFCD00213348 CAS nummer: 96478-09-0 EINECS: 424-240-4 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 96.0 to 100.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...