1-(2-Hydroxyethyl)-3-methylimidazolium chloride ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1-(2’-hydroxylethyl)-3-methylimidazolium chloride
H1254-5G
H1254-25G
TCIAH1254-5GEA
335
SEK
TCIAH1254-5G
TCIAH1254-25G
1-(2-Hydroxyethyl)-3-methylimidazolium chloride ≥98.0%
1-(2-Hydroxyethyl)-3-methylimidazolium chloride
Formel:
C₆H₁₁ClN₂O MW: 162,6173 g/mol |
MDL Number:
MFCD12761437 CAS nummer: 61755-34-8 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 82.0 to 87.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...