Harmaline ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 3,4-Dihydro-7-methoxy-1-methyl-beta-carboline, 3,4-Dihydro-7-methoxy-1-methyl-9H-pyrido[3,4-b]indole, Dihydroharmine

H1237-1G H1237-5G
TCIAH1237-1GEA 605 SEK
TCIAH1237-1G TCIAH1237-5G
Harmaline ≥98.0% (by HPLC, titration analysis)
Harmaline
Formel: C₁₃H₁₄N₂O
MW: 214,27 g/mol
Smältpunkt: 251°C
MDL Number: MFCD00004955
CAS nummer: 304-21-2
EINECS: 206-152-6
Merck Index: 14,04613

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR