6-Heptynenitrile ≥98.0%

Leverantör: TCI
Fara

H1214-25G H1214-5G
TCIAH1214-25GEA 6600 SEK
TCIAH1214-25G TCIAH1214-5G
6-Heptynenitrile ≥98.0%
6-Heptynenitrile
Formel: C₇H₉N
Kokpunkt: 93°C
CAS nummer: 15295-69-9
UN: 3276
ADR: 6.1,III

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR