(S)-2-Amino-6-guanidinohexanoic acid hydrochloride ≥97.0%

Leverantör: TCI EUROPE

Synonyms: H-HoArg-OH·HCl, L(+)-Homoarginine hydrochloride, H-HomoArg-OH.HCl, H-HomoArg-OH·HCl

H1172-1G H1172-5G
TCIAH1172-1GEA 430 SEK
TCIAH1172-1G TCIAH1172-5G
(S)-2-Amino-6-guanidinohexanoic acid hydrochloride ≥97.0%
(S)-2-Amino-6-guanidinohexanoic acid hydrochloride
Formel: C₇H₁₆N₄O₂·HCl
MW: 224,7 g/mol
Smältpunkt: 206°C
MDL Number: MFCD00012622
CAS nummer: 1483-01-8
EINECS: 216-045-6

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Specific rotation [a]20/D +16.0 to +20.0 deg (C=8,6N HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR