4-(2-Hydroxyethyl)thiomorpholine-1,1-dioxide ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Synonyms:
4-(2-Hydroxyethyl)thiomorpholine 1,1-Dioxide, 2-(1,1-Dioxothiomorpholino)ethanol
H1140-25G
H1140-5G
TCIAH1140-25GEA
4550
SEK
TCIAH1140-25G
TCIAH1140-5G
4-(2-Hydroxyethyl)thiomorpholine-1,1-dioxide ≥98.0% (by GC, titration analysis)
4-(2-Hydroxyethyl)thiomorpholine-1,1-dioxide
Formel:
C₆H₁₃NO₃S MW: 179,23 g/mol Kokpunkt: 197°C |
MDL Number:
MFCD00106415 CAS nummer: 26475-62-7 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 73.0 to 77.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...