4-Hydroxy-1,2,2,6,6-pentamethylpiperidine ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1,2,2,6,6-Pentamethylpiperidin-4-ol, 1,2,2,6,6-Pentamethyl-4-piperidinol

H1068-25G H1068-500G
TCIAH1068-25GEA 245 SEK
TCIAH1068-25G TCIAH1068-500G
4-Hydroxy-1,2,2,6,6-pentamethylpiperidine ≥98.0%
4-Hydroxy-1,2,2,6,6-pentamethylpiperidine
Formel: C₁₀H₂₁NO
MW: 171,28 g/mol
Kokpunkt: 120°C
MDL Number: MFCD00191220
CAS nummer: 2403-89-6
EINECS: 219-292-8

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 74.0 to 78.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR