Hexyl nitrite ≥95.0%
Leverantör: TCI EUROPE
|
Fara
|
H1066-25ML
H1066-500ML
TCIAH1066-25MLEA
485
SEK
TCIAH1066-25ML
TCIAH1066-500ML
Hexyl nitrite ≥95.0%
Hexyl nitrite
Formel:
C₆H₁₃NO₂ MW: 131,18 g/mol Kokpunkt: 52°C Storage Temperature: Kylskåp |
MDL Number:
MFCD06797110 CAS nummer: 638-51-7 UN: 1993 ADR: 3,II |
Specifikation testresultat
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...