Itaconic anhydride 98%
Leverantör: Thermo Scientific
Fara
Synonyms: Methylenesuccinic anhydride, 3-Methylenedihydrofuran-2,5-dione
412940250 412941000
ACRO412940250EA 1350 SEK
ACRO412940250 ACRO412941000
Itaconic anhydride 98%
Itaconic anhydride
Formel: C₅H₄O₃
MW: 112,08 g/mol
Kokpunkt: 239 °C (1013 hPa)
Smältpunkt: 66…70 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005530
CAS nummer: 2170-03-8
EINECS: 218-518-2
Order Now
Specifikation testresultat
Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 66 to 70 °C
Assay ≥97.5 %