4-(4-Hydroxy-3-methoxyphenyl)-3-buten-2-one ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Vanillylideneacetone, Dehydrozingerone, 4-(4-Hydroxy-3-methoxyphenyl)but-3-en-2-one

H0996-25G H0996-5G
TCIAH0996-25GEA 2600 SEK
TCIAH0996-25G TCIAH0996-5G
4-(4-Hydroxy-3-methoxyphenyl)-3-buten-2-one ≥98.0%
4-(4-Hydroxy-3-methoxyphenyl)-3-buten-2-one
Formel: C₁₁H₁₂O₃
MW: 192,21 g/mol
MDL Number: MFCD00012210
CAS nummer: 1080-12-2

Order Now

Specifikation testresultat

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 127.0 to 132.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR