9-(2-Hydroxyethyl)anthracene ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 2-(9-Anthryl)ethanol

H0970-1G H0970-5G
TCIAH0970-1GEA 2675 SEK
TCIAH0970-1G TCIAH0970-5G
9-(2-Hydroxyethyl)anthracene ≥98.0%
9-(2-Hydroxyethyl)anthracene
Formel: C₁₆H₁₄O
MW: 222,29 g/mol
MDL Number: MFCD01318099
CAS nummer: 54060-73-0

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 121.0 to 125.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR