4-(3-Hydroxyphenyl)piperidine ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3-(Piperidin-4-yl)phenol, 3-(4-Piperidyl)phenol

H0965-25G H0965-5G
TCIAH0965-25GEA 24400 SEK
TCIAH0965-25G TCIAH0965-5G
4-(3-Hydroxyphenyl)piperidine ≥98.0%
4-(3-Hydroxyphenyl)piperidine
Formel: C₁₁H₁₅NO
MW: 177,25 g/mol
MDL Number: MFCD02093474
CAS nummer: 110878-71-2

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR