Helicin ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
2-Formylphenyl beta-D-Glucopyranoside, 2-(((2S,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)benzaldehyde, Salicylaldehyde-beta-D-glucoside, 2-(beta-D-Glucopyranosyloxy)benzaldehyde
H0908-1G
H0908-5G
TCIAH0908-1GEA
655
SEK
TCIAH0908-1G
TCIAH0908-5G
Helicin ≥98.0%
Helicin
Formel:
C₁₃H₁₆O₇ MW: 284,26 g/mol |
MDL Number:
MFCD00006589 CAS nummer: 618-65-5 EINECS: 210-558-9 Merck Index: 14,04628 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Specific rotation [a]20/D | -59.0 to -64.0 deg (C=0.8, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...