2,3,6,7,10,11-Hexahydroxytriphenylene ≥95.0%

Leverantör: TCI
Varning

Synonyms: Triphenylene-2,3,6,7,10,11-hexaol

H0907-1G H0907-5G
TCIAH0907-1GEA 1525 SEK
TCIAH0907-1G TCIAH0907-5G
2,3,6,7,10,11-Hexahydroxytriphenylene ≥95.0%
2,3,6,7,10,11-Hexahydroxytriphenylene
Formel: C₁₈H₁₂O₆
MW: 324.29 g/mol
MDL Number: MFCD01321170
CAS nummer: 4877-80-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4af83611406ffd65fd04b99f74190c012094220&vid=6a80a2a627a2273a9dd7183839894a147b2741b8&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Red to Dark blue to Black powder to crystal
Purity (HPLC) min. 95.0 area%
Water max. 8.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR