2-Hydrazinylpyridine ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2(1H)-Pyridinone, hydrazone, 2-hydrazinylpyridine, 2-Hydrazinopyridine

H0888-10G H0888-25G
TCIAH0888-10GEA 1825 SEK
TCIAH0888-10G TCIAH0888-25G
2-Hydrazinylpyridine ≥98.0%
2-Hydrazinylpyridine
Formel: C₅H₇N₃
MW: 109,13 g/mol
Kokpunkt: 185°C
Storage Temperature: Kylskåp
MDL Number: MFCD00006249
CAS nummer: 4930-98-7
EINECS: 225-566-8

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystaline
Purity (GC) min. 98.0 %
Purity (Potassium iodate Method) min. 97.0 %
Melting point 44.0 to 48.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR