4-Hydroxy-4'-methoxybiphenyl ≥95.0%
Leverantör: TCI EUROPE
Synonyms:
4-Hydroxy-4'-methoxydiphenyl, 4'-Methoxy-[1,1'-biphenyl]-4-ol, 4'-Methoxybiphenyl-4-ol
H0819-1G
H0819-5G
TCIAH0819-1GEA
1100
SEK
TCIAH0819-1G
TCIAH0819-5G
4-Hydroxy-4'-methoxybiphenyl ≥95.0%
4-Hydroxy-4'-methoxybiphenyl
Formel:
C₁₃H₁₂O₂ MW: 200,24 g/mol |
MDL Number:
MFCD00191514 CAS nummer: 16881-71-3 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 180.0 to 185.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...