(+/-)-Hydrobenzoin ≥98.0%

Leverantör: TCI EUROPE

Synonyms: (+/-)-1,2-Diphenyl-1,2-ethanediol

H0815-1G H0815-5G
TCIAH0815-1GEA 815 SEK
TCIAH0815-1G TCIAH0815-5G
(+/-)-Hydrobenzoin ≥98.0%
(+/-)-Hydrobenzoin
Formel: C₁₄H₁₄O₂
MW: 214,26 g/mol
MDL Number: MFCD00136059
CAS nummer: 655-48-1
Merck Index: 14,04777

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light red powder to crystaline
Purity (GC) min. 98.0 %
Melting point 120.0 to 125.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR