Dimethyl-trans-3-hexenedioate ≥98.0%
Leverantör: TCI EUROPE
Synonyms: trans-3-Hexenedioic Acid Dimethyl Ester
H0776-25G H0776-5G
TCIAH0776-25GEA 3225 SEK
TCIAH0776-25G TCIAH0776-5G
Dimethyl-trans-3-hexenedioate ≥98.0%
Dimethyl-trans-3-hexenedioate
Formel: C₈H₁₂O₄
MW: 172,18 g/mol
Kokpunkt: 125°C
MDL Number: MFCD00145425
CAS nummer: 25126-93-6
Order Now
Specifikation testresultat
Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %