4-Hydroxy-4'-nitrobiphenyl ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 4'-Nitrobiphenyl-4-ol, 4'-Nitro-[1,1'-biphenyl]-4-ol

H0747-1G H0747-5G H0747-25G
TCIAH0747-1GEA 695 SEK
TCIAH0747-1G TCIAH0747-5G TCIAH0747-25G
4-Hydroxy-4'-nitrobiphenyl ≥98.0%
4-Hydroxy-4'-nitrobiphenyl
Formel: C₁₂H₉NO₃
MW: 215,21 g/mol
MDL Number: MFCD00047742
CAS nummer: 3916-44-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb58d71ef6541b3b328758d1d78b46a7a777b226&vid=bb58d71ef6541b3b328758d1d78b46a7a777b226&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 205.0 to 208.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR