Methyl-4-(2-hydroxyethoxy)salicylate ≥98.0%
Leverantör: TCI
H0727-1G
H0727-5G
TCIAH0727-1GEA
685
SEK
TCIAH0727-1G
TCIAH0727-5G
Methyl-4-(2-hydroxyethoxy)salicylate ≥98.0%
Methyl-4-(2-hydroxyethoxy)salicylate
|
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...