4-Hydroxyindanone ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 4-HYDROXY-2,3-DIHYDRO-1H-INDEN-1-ONE, 4-hydroxyindan-1-one, 4-Hydroxy-1-indanone

H0723-25G H0723-5G
TCIAH0723-25GEA 37750 SEK
TCIAH0723-25G TCIAH0723-5G
4-Hydroxyindanone ≥98.0% (by GC, titration analysis)
4-Hydroxyindanone
Formel: C₉H₈O₂
MW: 148,161 g/mol
MDL Number: MFCD00143330
CAS nummer: 40731-98-4

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR