4-Hexyloxyphthalonitrile ≥95.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1,2-Dicyano-4-hexyloxybenzene

H0691-1G H0691-5G
TCIAH0691-1GEA 895 SEK
TCIAH0691-1G TCIAH0691-5G
4-Hexyloxyphthalonitrile ≥95.0%
4-Hexyloxyphthalonitrile
Formel: C₁₄H₁₆N₂O
MW: 228,30 g/mol
MDL Number: MFCD00059077
CAS nummer: 104949-82-8

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR