N-(Hydroxymethyl)phthalimide ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
Synonyms:
2-(Hydroxymethyl)isoindoline-1,3-dione
H0648-25G
H0648-250G
TCIAH0648-25GEA
215
SEK
TCIAH0648-25G
TCIAH0648-250G
N-(Hydroxymethyl)phthalimide ≥98.0% (by HPLC, total nitrogen)
N-(Hydroxymethyl)phthalimide
Formel:
C₉H₇NO₃ MW: 177,16 g/mol |
MDL Number:
MFCD00005899 CAS nummer: 118-29-6 EINECS: 204-241-4 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 143.0 to 148.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...