Ishikawa's reagent ≥92.0% fluorinating reagent

Leverantör: TCI
Fara

H0598-25G H0598-100G
TCIAH0598-25GEA 1075 SEK
TCIAH0598-25G TCIAH0598-100G
Ishikawa's reagent ≥92.0% fluorinating reagent
Ishikawa's reagent
Formel: C₇H₁₁F₆N
Kokpunkt: 54°C
Flampunkt: 40°C
MDL Number: MFCD00060264
CAS nummer: 309-88-6
UN: 2920
ADR: 8,II
Merck Index: 14,05108
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9a5186904a774484d51d5394744d38a6a97aa4e&vid=a9a5186904a774484d51d5394744d38a6a97aa4e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Green clear liquid
Purity (Neutralization titration) min. 92.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR